Learn More
Bergen Industrial Supply 1.5IN SILICON SCREEN GASKET325
Catalog No. NC2131780
Encompass_Preferred
RUBBERFAB TECHNOLOGIES 1.50" SCREENEDGASKET PLATINUM-CURED SILICONE, 316SS325-MESH, USP-CLASS-VI W/CERTSVendor Part number 40RXPXS-150-325SS
This item is not returnable. View return policy
Catalog No. NC2131780 Supplier Bergen Industrial Supply Supplier No. 40RXPXS150325SS
RUBBERFAB TECHNOLOGIES 1.50" SCREENEDGASKET PLATINUM-CURED SILICONE, 316SS325-MESH, USP-CLASS-VI W/CERTSVendor Part number 40RXPXS-150-325SS