Learn More
Ace Glass, Inc. Heating tape, 5/8in, 5ft length, 145w at 115v max, Glas-Col; 103A FET0.65
SDP
Catalog No. 50977375
Encompass
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 5 foot length. Can be operated directly through a temperature control at normal outlet voltage. 145w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.
Catalog No. 50-977-375 Supplier Ace Glass, Inc. Supplier No. 1206302
5/8 Glas-Col heating tape allows the use of 115v without the need of a reducing voltage transformer. 5 foot length. Can be operated directly through a temperature control at normal outlet voltage. 145w at 115v max. Operating temperature to 249 degrees C. Both power supply connections are made at one end of the tape. Supplied with a separable 1.2m extension cord with two-blade cup.