Learn More
Analytical Sales & Services 1.5mL 96-Well Opaque Flexi-Tier™ Block System
Supplier:  Analytical Sales & Services 96FP15
Encompass
1.5mL 96-Well Opaque Flexi-Tier System\n\nIncludes:\n• 1.5mL Conical Glass Inserts\n• 2 Spacers\n• Base Plate\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-06747