Learn More
Analytical Sales & Services 1.5mL 96-Well Opaque Flexi-Tier™ Block System
Catalog No. 5000804215
Encompass
1.5mL 96-Well Opaque Flexi-Tier Block System\n\nIncludes:\n• 1.5mL Flat Bottom Glass Inserts\n• Base Plate\n• 2 Spacers\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH
Catalog No. 50-008-04215 Supplier Analytical Sales & Services Supplier No. 96BB15
1.5mL 96-Well Opaque Flexi-Tier Block System\n\nIncludes:\n• 1.5mL Flat Bottom Glass Inserts\n• Base Plate\n• 2 Spacers\n• Pre-Scored PTFE/Silicone Flat Liner\n• Cover\n\nQty/Pkg: EACH