Learn More
Analytical Sales & Services 1.5mL Aluminum Block System, Solid Base with Flat Bottom Inserts
Catalog No. 5000804590
Encompass
1.5mL Aluminum Block System with Solid Base. Includes: Aluminum Base, Cover, Screws, Molded Silicone Liner and 1.5mL Clear Glass Flat Bottom Inserts Preassembled\n\nQty/Pkg: EACH
Catalog No. 50-008-04590 Supplier Analytical Sales & Services Supplier No. 96F115
1.5mL Aluminum Block System with Solid Base. Includes: Aluminum Base, Cover, Screws, Molded Silicone Liner and 1.5mL Clear Glass Flat Bottom Inserts Preassembled\n\nQty/Pkg: EACH