Learn More
Diagenode Inc 1.5ML BIORUPTR MICROT W/CP 300
Catalog No. NC0605655
Encompass
The 1.5 Ml Bioruptor Microtubes For Chromatin Shearing Strongly Improve Chromatin Sonication Efficiency. These Tubes Have Been Thoroughly Validated For Use On The Bioruptor Pico (B01060001) And Are Strongly Recommended For Chromatin Shearing Applications Using This Instrument. Recommended Sample Volume: 100 Μl - 300 Μl.
Catalog No. NC0605655 Supplier Diagenode Inc Supplier No. C30010016
The 1.5 Ml Bioruptor Microtubes For Chromatin Shearing Strongly Improve Chromatin Sonication Efficiency. These Tubes Have Been Thoroughly Validated For Use On The Bioruptor Pico (B01060001) And Are Strongly Recommended For Chromatin Shearing Applications Using This Instrument. Recommended Sample Volume: 100 Μl - 300 Μl.