Learn More
Chemglass Life Sciences 1.5mL Clear Graduated High Recovery Vial with 13-425 Black Phenolic Open Top Cap and Red PTFE/Silicone Septa, 13mm OD x 43.5mm Height
SDP
Catalog No. 5000980623
Encompass_Preferred
Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa.
Catalog No. 50-009-80623 Supplier Chemglass Life Sciences Supplier No. CG4913V015
Vials are manufactured from Type 1, Class A Borosilicate glass and come with an open-top cap with PTFE Silicone septa.