Learn More
CELLTREAT Scientific Products 1.5mL Black Micro Centrifuge Tube, Sterile
SDP
Catalog No. 50403867
Encompass_Preferred
CELLTREAT Scientific Products 1.5mL Black Micro Centrifuge Tube. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Black tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil. RNase/DNase free and non-pyrogenic. Max force (RCF) g 20000. Temperature range of -80¿C to 120¿C. Every case is printed with the lot number for quality traceability. Packaged sterile, 500/resealable bag, 1000/case.
Catalog No. 50-403-867 Supplier CELLTREAT Scientific Products Supplier No. 229437
CELLTREAT Scientific Products 1.5mL Black Micro Centrifuge Tube. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Black tubes protect light sensitive compounds and eliminate the need to wrap tubes with aluminum foil. RNase/DNase free and non-pyrogenic. Max force (RCF) g 20000. Temperature range of -80¿C to 120¿C. Every case is printed with the lot number for quality traceability. Packaged sterile, 500/resealable bag, 1000/case.