Learn More
USA Scientific Inc 1.5 mL conical screw cap tube, violet cap
Catalog No. 507541457
Encompass_Preferred
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural with violet cap, 500/pack
Catalog No. 50-754-1457 Supplier USA Scientific Inc Supplier No. 14158705
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural with violet cap, 500/pack