Learn More
USA Scientific Inc 1.5 mL conical screw cap tube, yellow cap
Catalog No. 507541458
Encompass_Preferred
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural with yellow cap, 500/pack
Catalog No. 50-754-1458 Supplier USA Scientific Inc Supplier No. 14158706
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural with yellow cap, 500/pack