Learn More
USA Scientific Inc 1.5mL conical tube, mixed
Catalog No. 502745033
Encompass
1.5 mL conical base microcentrifuge tube, graduated, mixed colors, with integrated TPE cap, 500/pack
Catalog No. 50-274-5033 Supplier USA Scientific Inc Supplier No. 14158399
1.5 mL conical base microcentrifuge tube, graduated, mixed colors, with integrated TPE cap, 500/pack