Learn More
Chemglass Life Sciences Chemglass 1.5 mL Conical Sample Vials, External Thread, Natural Caps, Sterile (CGN-4764-004N) - 50/pk
SDP
Catalog No. 502733359
Encompass_Preferred
Sterile 1.5 mL conical sample vials with external thread and natural-color caps. Each vial includes a sealing ring and is supplied in nest-format packaging. Produced in a 100,000-level clean workshop and sterilized by electron beam (SAL 10^-6), these vials are suitable for laboratory sampling and storage.
  • Capacity: 1.5 mL.
  • External thread with sealing ring.
  • Natural-color caps.
  • Sterile - electron beam sterilization (SAL = 10^-6).
  • Packaging: 50 per pack, 500 per box, 2,000 per case.
  • Produced in 100,000-level clean workshop.
  • DNAse-free, RNAse-free, pyrogen-free, endotoxin-free.
Catalog No. 50-273-3359 Supplier Chemglass Life Sciences Supplier No. CGN4764004N
Add to Cart
Add to Cart
Sterile 1.5 mL conical sample vials with external thread and natural-color caps. Each vial includes a sealing ring and is supplied in nest-format packaging. Produced in a 100,000-level clean workshop and sterilized by electron beam (SAL 10^-6), these vials are suitable for laboratory sampling and storage.
  • Capacity: 1.5 mL.
  • External thread with sealing ring.
  • Natural-color caps.
  • Sterile - electron beam sterilization (SAL = 10^-6).
  • Packaging: 50 per pack, 500 per box, 2,000 per case.
  • Produced in 100,000-level clean workshop.
  • DNAse-free, RNAse-free, pyrogen-free, endotoxin-free.