Learn More
Chemglass Life Sciences Chemglass 1.5 mL conical sample vial, external thread with hinged sealing cap, natural color, 50/pack
SDP
Catalog No. 502733355
Encompass_Preferred
Chemglass 1.5 mL conical sample vials are sterile, type I borosilicate glass vials designed for sample storage and analytical workflows. They feature an external thread and a hinged cap with a sealing ring for secure closure, and are supplied nested for convenient handling.
  • 1.5 mL capacity.
  • Type I borosilicate glass.
  • Natural color cap.
  • External thread.
  • Hinged cap with sealing ring.
  • Sterile.
  • Packaged 50 per pack, 500 per box, 2,000 per case.
Catalog No. 50-273-3355 Supplier Chemglass Life Sciences Supplier No. CGN4764014N
Add to Cart
Add to Cart
Chemglass 1.5 mL conical sample vials are sterile, type I borosilicate glass vials designed for sample storage and analytical workflows. They feature an external thread and a hinged cap with a sealing ring for secure closure, and are supplied nested for convenient handling.
  • 1.5 mL capacity.
  • Type I borosilicate glass.
  • Natural color cap.
  • External thread.
  • Hinged cap with sealing ring.
  • Sterile.
  • Packaged 50 per pack, 500 per box, 2,000 per case.