Learn More
Alkali Scientific 1.5mL Crimp High Recovery Vial Clear Glass Flat Base Wide Opening, 100/Pk
SDP
Catalog No. 5000661929
Encompass_Preferred
The Alkali Scientific™ 1.5 mL Crimp High Recovery Vial is engineered for maximum sample recovery and dependable performance in analytical and laboratory applications. Manufactured from high quality clear glass, this vial offers excellent chemical resistance while allowing full visibility of sample contents during preparation and analysis. The high recovery design reduces residual sample loss, helping ensure more accurate results in sensitive procedures. A flat base provides stability during handling and instrument use, while the wide opening allows easy filling and cleaning. The crimp compatible finish enables secure sealing with standard crimp caps for reliable storage and transport. Supplied in convenient packs of 100, these vials are ideal for chromatography, sample storage, and high throughput laboratory workflows.
Catalog No. 50-006-61929 Supplier Alkali Scientific Supplier No. HRV01.5CT
The Alkali Scientific™ 1.5 mL Crimp High Recovery Vial is engineered for maximum sample recovery and dependable performance in analytical and laboratory applications. Manufactured from high quality clear glass, this vial offers excellent chemical resistance while allowing full visibility of sample contents during preparation and analysis. The high recovery design reduces residual sample loss, helping ensure more accurate results in sensitive procedures. A flat base provides stability during handling and instrument use, while the wide opening allows easy filling and cleaning. The crimp compatible finish enables secure sealing with standard crimp caps for reliable storage and transport. Supplied in convenient packs of 100, these vials are ideal for chromatography, sample storage, and high throughput laboratory workflows.