Learn More
Azzota Corp Disposable Uv-Vis Cuvettes - 1.5Ml X 100 Square Cuvettes 10mm X 4mm X 45mm Methacrylate Pathlength: 10mm 1.5Ml Volume Wavelength Range: 285 To 800Nm (Uv-Vis) 100 Pack
Catalog No. 507532901
Encompass_Preferred
Disposable UV-VIS Cuvettes - 1.5ml x 100 Square Cuvettes 10mm x 4mm x 45mm Methacrylate Pathlength: 10mm 1.5ml volume Wavelength Range: 285 to 800nm (UV-Vis) 100 pack
Catalog No. 50-753-2901 Supplier Azzota Corp Supplier No. A908
Disposable UV-VIS Cuvettes - 1.5ml x 100 Square Cuvettes 10mm x 4mm x 45mm Methacrylate Pathlength: 10mm 1.5ml volume Wavelength Range: 285 to 800nm (UV-Vis) 100 pack