Learn More
USA Scientific Inc 1.5 mL low adhesion microcentrifuge tube, mixed
Catalog No. 507541451
Encompass
1.5 mL low adhesion polypropylene microcentrifuge tube, mixed, graduated, 500/bag
Catalog No. 50-754-1451 Supplier USA Scientific Inc Supplier No. 14152608
1.5 mL low adhesion polypropylene microcentrifuge tube, mixed, graduated, 500/bag