Learn More
Alkali Scientific 1.5mL High Recovery Vial Glass 12x32mm Flat Base 9 425 Screw Thread. 100/Pk, Clear
SDP
Supplier:  Alkali Scientific HRV01.5
Encompass_Preferred
The Alkali Scientific™ 1.5 mL High Recovery Vial is designed for maximum sample retrieval and reliable performance in laboratory and analytical applications. Manufactured from high quality clear glass, this vial provides excellent chemical resistance and visibility of contents. The high recovery design minimizes residual sample loss, making it ideal for low volume or valuable samples. The flat base ensures stability during handling and autosampler use, while the 9 425 screw thread finish is compatible with standard caps and closures. Supplied in packs of 100, these vials are perfect for routine analysis, sample storage, and high throughput laboratory workflows. Key Features 1.5 mL capacity for high recovery sample applications High recovery design minimizes residual sample loss Clear glass construction for excellent chemical resistance and sample visibility Flat base provides stability during handling and autosampler use 12 × 32 mm dimensions suitable for labor
Catalog No. 50-006-61932