Learn More
Analytical Sales & Services 1.5mL Low Adhesion Screw Top Micro Tube without Skirt
Catalog No. 5000804275
Encompass
1.5mL Low Adhesion Screw Top Micro Tube without Skirt\nCS 1000, Polypropylene\nTemperature Range: -196 ¡C to +121 ¡C\nCertified RNase-, DNase-, Pyrogen- and DNA-free.\n\nQty/Pkg: 1000
Catalog No. 50-008-04275 Supplier Analytical Sales & Services Supplier No. 16606
1.5mL Low Adhesion Screw Top Micro Tube without Skirt\nCS 1000, Polypropylene\nTemperature Range: -196 ¡C to +121 ¡C\nCertified RNase-, DNase-, Pyrogen- and DNA-free.\n\nQty/Pkg: 1000