Learn More
Chemglass Life Sciences 1.5mL Micro Centrifuge Tube, Clear, Conical, Lock Cap, Sterile
SDP
Catalog No. 501897711
Encompass_Preferred
¿ USP VI grade polypropylene material, free from heavy metal ¿ RCF: 30,000 x g ¿ Endotoxin level: <0.1EU ¿ Clear graduations and frosted marking area ¿ Special Safe-lock cap design provides secure lid sealing ¿ Easy opening and closing ¿ Temperature range: -80? to +121? ¿ DNase/RNase-freeTubes are autoclavable at 121¿C (250¿F), under 15psi for 10¿15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.
Catalog No. 50-189-7711 Supplier Chemglass Life Sciences Supplier No. CGN430015N
¿ USP VI grade polypropylene material, free from heavy metal ¿ RCF: 30,000 x g ¿ Endotoxin level: <0.1EU ¿ Clear graduations and frosted marking area ¿ Special Safe-lock cap design provides secure lid sealing ¿ Easy opening and closing ¿ Temperature range: -80? to +121? ¿ DNase/RNase-freeTubes are autoclavable at 121¿C (250¿F), under 15psi for 10¿15 minutes with drying. Remove from autoclave immediately. Excessive exposure to autoclaving heat may cause warping or decreased structural integrity.