Learn More
CELLTREAT Scientific Products 1.5mL Micro Centrifuge Tube, Black, Non-sterile
SDP
Catalog No. 5000144342
Encompass_Preferred
All tubes have a frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handColor tubes allow for quick sample identification, or to customize workflows
Catalog No. 50-001-44342 Supplier CELLTREAT Scientific Products Supplier No. 230435
All tubes have a frosted marking spot and engraved graduationsAttached flat top cap is easy to open and close with one handColor tubes allow for quick sample identification, or to customize workflows