Learn More
CELLTREAT Scientific Products 1.5mL Micro Centrifuge Tube, Sterile
SDP
Supplier:  CELLTREAT Scientific Products 229442
Encompass_Preferred
CELLTREAT Scientific Products 1.5mL Micro Centrifuge Tube. Tubes have frosted marking spot and engraved graduations. Attached flat top cap is easy to open and close with one hand. Temperature range of -80¿C to 120¿C. Rnase/Dnase free and non-pyrogenic. Max force (RCF) g 20000. Every case is printed with the lot number for quality traceability. Packaged sterile, 500/re-sealable bag, 1000/case.
Catalog No. 50-202-024