Learn More
Midwest Scientific 1.5ML MICRO TB NS RAINBOW 500
SDP
Catalog No. NC1845976
Encompass
PR1MA Microcentrifuge Tubes; 1.5ml; 500/pk; non-sterile, Cap Style: Snap small cap boil proof;Temp Range:-80C-121C; Assorted colors; Midwest Scientific mfg# PR-MCT15-RB
Catalog No. NC1845976 Supplier Midwest Scientific Supplier No. PRMCT15RB
PR1MA Microcentrifuge Tubes; 1.5ml; 500/pk; non-sterile, Cap Style: Snap small cap boil proof;Temp Range:-80C-121C; Assorted colors; Midwest Scientific mfg# PR-MCT15-RB