Learn More
USA Scientific Inc 1.5 mL copolymer microcentrifuge tube, orange
Catalog No. NC9439727
Encompass_Preferred
1.5 mL copolymer polypropylene graduated microcentrifuge tube, orange, 500/bag
This item is not returnable. View return policy
Catalog No. NC9439727 Supplier USA Scientific Inc Supplier No. 14152503
1.5 mL copolymer polypropylene graduated microcentrifuge tube, orange, 500/bag