Learn More
Alkali Scientific Microcentrifuge Tube, 1.5mL, Clear, Conical, Lock Cap, Non-Sterile, 500/pk, 5000/cs
SDP
Catalog No. 502335756
Encompass_Preferred
USP VI grade PP material, free from heavy metal. RCF: 30000xg. Endotoxin: 0.1EU. They have clear graduations and frosted marking area, and a special cap design to insert needles. Easy opening and closing. They can withstand temperature ranges of -80℃ to +121℃. DNase, RNase, and pyrogen-free. Autoclave at 121℃/15 psi.
Catalog No. 50-233-5756 Supplier Alkali Scientific Supplier No. CN3015
USP VI grade PP material, free from heavy metal. RCF: 30000xg. Endotoxin: 0.1EU. They have clear graduations and frosted marking area, and a special cap design to insert needles. Easy opening and closing. They can withstand temperature ranges of -80℃ to +121℃. DNase, RNase, and pyrogen-free. Autoclave at 121℃/15 psi.