Learn More
Alkali Scientific Microcentrifuge Tube, 1.5mL, Clear, Sterile, Conical, Lock Cap, 50/bag, 400/pk, 4000/cs
SDP
Catalog No. 502335987
Encompass_Preferred
These centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. USP VI grade polypropylene material, free from heavy metal. Special safe-lock cap design provides secure lid sealing and makes them easy to open and close. They have clear graduations and frosted marking area. RNase free, endotoxin level less than 0.1EU. Autoclavable
Catalog No. 50-233-5987 Supplier Alkali Scientific Supplier No. CN3016
These centrifuge tubes could be commonly used to collect cells and separate biomacromolecules by centrifugation in biochemistry and other biological fields. They can also be used to store photosensitive samples and special chemiluminescence reagents. USP VI grade polypropylene material, free from heavy metal. Special safe-lock cap design provides secure lid sealing and makes them easy to open and close. They have clear graduations and frosted marking area. RNase free, endotoxin level less than 0.1EU. Autoclavable