Learn More
MTC Bio ClearSeal™ screw-cap microtubes, 1.5mL , w/ O-ring & caps, sterile, printed graduations, self-standing, 20 resealable bags of 50 tubes, 1000/cs
Catalog No. NC2017388
Encompass_Preferred
ClearSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and even a loop-cap option for the 2.0mL. Made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg*.
Catalog No. NC2017388 Supplier MTC Bio Supplier No. C3215SG
ClearSeal™ sterile screw-cap microtubes come in a variety of sizes, cap configurations, graduation options, and even a loop-cap option for the 2.0mL. Made from ultra-clear polypropylene, these screw-cap microtubes include assembled caps with silicone O-rings that assure a leakproof seal from -90°C to +121°C. All ClearSeal™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg*.