Learn More
Genesee Scientific Corporation Clear Large Cap Microtube, 1.7mL
SDP
Catalog No. NC9259991
Encompass
Microtube, 1.7mL, Clear Large Cap, PP, Boil-Resistant, Leakproof, Certified RNase, DNase and Pyrogen Safe, Engineered to Close with a Snap and Stay Closed During Boiling and Centrifugation, Can be Used in Any Centrifuge Up to 20, 000 x g
Catalog No. NC9259991 Supplier Genesee Scientific Corporation Supplier No. 22281
Microtube, 1.7mL, Clear Large Cap, PP, Boil-Resistant, Leakproof, Certified RNase, DNase and Pyrogen Safe, Engineered to Close with a Snap and Stay Closed During Boiling and Centrifugation, Can be Used in Any Centrifuge Up to 20, 000 x g