Learn More
Microsolv Technology Corporation 1.5Ml Mr Vials W/Snaptop Deac
Catalog No. 5001452311
Encompass_Preferred
Vials, Snap Top, Glass. Clear, 1.8ml, Max Recovery™. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.
Catalog No. 50-014-52311 Supplier Microsolv Technology Corporation Supplier No. 9512C0DCV
Vials, Snap Top, Glass. Clear, 1.8ml, Max Recovery™. Deactivated. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. AQ Brand. 100/PK.