Learn More
Sarstedt Inc 1.5ML SCREW CAP MCRTUBE 500/PK
Supplier:  Sarstedt Inc 72.607.772
Encompass_Preferred
Screw Cap Micro Tube, 1.5 ml, PP, without smear edge, with knurls, without closure, no print, sterile, 500 pcs./double bag.
Catalog No. NC1532983