Learn More
PERFECTOR SCIENTIFIC Polystyrene cuvettes 1.5ml semi-micro. 500/pk
Catalog No. NC0177642
Encompass
All cuvettes have a 10mm path length and are 45mm high. The internal width of the semi-micro cuvette is 4mm. Fluorometer cuvettes have 4 sides clear. Packed in 5 styrofoam trays with each tray holding 100 cuvettes. The trays have a tight fitting styrofoam lid which protects the optical surfaces and keeps the cuvettes clean. 500/pk
Catalog No. NC0177642 Supplier PERFECTOR SCIENTIFIC Supplier No. 9003
All cuvettes have a 10mm path length and are 45mm high. The internal width of the semi-micro cuvette is 4mm. Fluorometer cuvettes have 4 sides clear. Packed in 5 styrofoam trays with each tray holding 100 cuvettes. The trays have a tight fitting styrofoam lid which protects the optical surfaces and keeps the cuvettes clean. 500/pk