Learn More
Andwin Scientific 1.5ML SPERSPN TUBE MICRO 500PK
SDP
Catalog No. NC9329347
Encompass_Preferred
The 1.5 mL SuperSpin™ Microcentrifuge Tubes are ideal for vortexing and shaking tables. This product is able to endure centrifugations spins up to 35,000 g, which is twice the g-force most other microcentrifuge tubes can tolerate. The tubes are also
Catalog No. NC9329347 Supplier Andwin Scientific Supplier No. 20170038
The 1.5 mL SuperSpin™ Microcentrifuge Tubes are ideal for vortexing and shaking tables. This product is able to endure centrifugations spins up to 35,000 g, which is twice the g-force most other microcentrifuge tubes can tolerate. The tubes are also