Learn More
BioSpec Products 1.5ML SS MICRO VLS WCAP 100PK
SDP
Catalog No. NC0348280
Encompass_Preferred
Stainless Steel Microvials have the same outer dimensions and weight as standard 2 ml microvials. They can be sealed with polyethylene flange caps, silicone rubber stopper caps or crimped septa caps.
Catalog No. NC0348280 Supplier BioSpec Products Supplier No. 2007
Stainless Steel Microvials have the same outer dimensions and weight as standard 2 ml microvials. They can be sealed with polyethylene flange caps, silicone rubber stopper caps or crimped septa caps.