Learn More
USA Scientific Inc 1.5mL ss screw cap tube
Supplier:  USA Scientific Inc 14159301
Encompass
1.5 mL self-standing microcentrifuge tube, blue, with integrated TPE cap, 500/pack
Catalog No. 50-274-5549