Learn More
Andwin Scientific 1.5ML STERIL CON TUBES 500PK
SDP
Catalog No. NC0485704
Encompass_Preferred
Screw-Cap Microcentrifuge Tubes DNase and RNase free. With attached purple caps. Cap includes silicone O-ring. 50 tubes with attached caps per sterile bag 10 sterile bags per pack. Autoclavable and freezable to -90[degrees]C.
Catalog No. NC0485704 Supplier Andwin Scientific Supplier No. 89004290PK
Screw-Cap Microcentrifuge Tubes DNase and RNase free. With attached purple caps. Cap includes silicone O-ring. 50 tubes with attached caps per sterile bag 10 sterile bags per pack. Autoclavable and freezable to -90[degrees]C.