Learn More
Alkali Scientific 1.5mL Sterile Microcentrifuge Tubes w/O Ring & attached Screw Caps, Printed Graduations, Self Standing, 50/Bag, 1000/Case
SDP
Catalog No. 5000663152
Encompass_Preferred
Ultra clear polypropylene Packaged in resealable bags of 50 Leak proof O ring seal Sterile, RNase, DNase & Pyrogen free White printed graduations CellPro™ screw cap microtubes come sterile, in a variety of s, cap configurations, graduation options, and even a loop cap option for the 2.0mL version. Made from ultra clear polypropylene, these screw cap microtubes include assembled caps with silicone O rings that assure a leakproof seal from 90°C to +121°C. All CellPro™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.
Catalog No. 50-006-63152 Supplier Alkali Scientific Supplier No. MT3215SG
Ultra clear polypropylene Packaged in resealable bags of 50 Leak proof O ring seal Sterile, RNase, DNase & Pyrogen free White printed graduations CellPro™ screw cap microtubes come sterile, in a variety of s, cap configurations, graduation options, and even a loop cap option for the 2.0mL version. Made from ultra clear polypropylene, these screw cap microtubes include assembled caps with silicone O rings that assure a leakproof seal from 90°C to +121°C. All CellPro™ screw cap microtubes are sterile, RNase, DNase, & pyrogen free and may be centrifuged at up to 22,000xg*. *Refers to liquids with a specific gravity of 1.0, using appropriate rotors, cushions, and adapters.