Learn More
Alkali Scientific ASI™ 1.5mL Strip Tube, PP, Clear, 800/Cs, Sterile
SDP
Catalog No. 503029087
Encompass_Preferred
ASI™1.5ml Strip Tube, provided as strips of six microcentrifuge tubes physically connected in a row, is ideal for any large-scale experiments. The strips can be cut into individual tubes and are universally applicable to centrifugation.  1.5ml strip tube may restrictedly be compatible to rotors, depending on the types of rotors being used for centrifugation. Compatibility test, before proceeding experiment, is highly recommended. (Compatible to most 24-, 30-hole rotors, and few 18-hole rotors) ▪ Smooth opening and tight closing▪ Imprinted external graduations with marking area▪ Central part in cap enough to be penetrated by syringe needle▪ Autoclavable▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free
Catalog No. 50-302-9087 Supplier Alkali Scientific Supplier No. PC60615
ASI™1.5ml Strip Tube, provided as strips of six microcentrifuge tubes physically connected in a row, is ideal for any large-scale experiments. The strips can be cut into individual tubes and are universally applicable to centrifugation.  1.5ml strip tube may restrictedly be compatible to rotors, depending on the types of rotors being used for centrifugation. Compatibility test, before proceeding experiment, is highly recommended. (Compatible to most 24-, 30-hole rotors, and few 18-hole rotors) ▪ Smooth opening and tight closing▪ Imprinted external graduations with marking area▪ Central part in cap enough to be penetrated by syringe needle▪ Autoclavable▪ Non – pyrogenic▪ Non – cytotoxic▪ DNase / RNase – free▪ Human DNA – free