Learn More
Andwin Scientific 1-5ML TIPS CLEAR 250/PK
SDP
Catalog No. NC9902821
Encompass_Preferred
The macro pipet tips are compatible to use with a wide variety of macro pipettors, including the ones manufactured by Biohit, GIlson, Nichiryo, HTL, and Labnet. The tips have a volume capacity of 10005000 L and are nonsterile. It is sold in a pack of 250.
Catalog No. NC9902821 Supplier Andwin Scientific Supplier No. 89087530
The macro pipet tips are compatible to use with a wide variety of macro pipettors, including the ones manufactured by Biohit, GIlson, Nichiryo, HTL, and Labnet. The tips have a volume capacity of 10005000 L and are nonsterile. It is sold in a pack of 250.