Learn More
USA Scientific Inc 1.5mL v-bot screw cap tube
Supplier:  USA Scientific Inc 14158304
Encompass
1.5 mL conical base microcentrifuge tube, graduated, red, with integrated TPE cap, 500/pack
Catalog No. 50-274-5145