Learn More
USA Scientific Inc 1.5mL v-bot screw cap tube
Supplier:  USA Scientific Inc 14158300
Encompass
1.5 mL conical base screw cap microcentrifuge tube, graduated, natural, with integrated TPE natural color cap, 500/pack
Catalog No. 50-274-4823