Learn More
Chemglass Life Sciences Vials Complete: 1.5mL, 9mm Screw Thread, 12mm OD x 32mm Height
SDP
Catalog No. 50872792
Encompass_Preferred
Vials Complete: 1.5mL, 9mm Screw Thread, 12mm OD x 32mm Height. These CD™ vials have a unique Patented* conical interior bottom permitting the contents to completely drain to the center bottom of the vial. Maximum extraction of the contents can be accomplished using a syringe or micropipette. Available complete with open top screw caps having a PTFE faced silicone septa or vials only. The 1.5mL has a bottom as shown in photo, while the 40mL has a bottom as illustrated in the line art.PLEASE NOTE: • VIALS AND CAPS ARE SHIPPED TOGETHER, BUT NOT ASSEMBLED.• VIALS ARE AVAILABLE IN AMBER FOR LIGHT SENSITIVE MATERIALS. PLEASE CONTACT US FOR DETAILS.*Patent Number D454,397 S
Catalog No. 50-872-792 Supplier Chemglass Life Sciences Supplier No. CG491501
Vials Complete: 1.5mL, 9mm Screw Thread, 12mm OD x 32mm Height. These CD™ vials have a unique Patented* conical interior bottom permitting the contents to completely drain to the center bottom of the vial. Maximum extraction of the contents can be accomplished using a syringe or micropipette. Available complete with open top screw caps having a PTFE faced silicone septa or vials only. The 1.5mL has a bottom as shown in photo, while the 40mL has a bottom as illustrated in the line art.PLEASE NOTE: • VIALS AND CAPS ARE SHIPPED TOGETHER, BUT NOT ASSEMBLED.• VIALS ARE AVAILABLE IN AMBER FOR LIGHT SENSITIVE MATERIALS. PLEASE CONTACT US FOR DETAILS.*Patent Number D454,397 S