Learn More
Sarstedt Inc 1.5ml SC Micro Tube DNA LB
Catalog No. NC1544083 Shop All Sarstedt Inc. Products
Encompass_Preferred
Screw Cap Micro Tube DNA Low Binding, 1.5 ml, PP, with skirted base, with knurls, with assembled cap, with printed writing space and graduation, certified PCR Performance Tested (free of DNase/RNase, DNA and PCR inhibitors), 100 pcs./bag
This item is not returnable. View return policy
Catalog No. NC1544083 Supplier Sarstedt Inc Supplier No. 72.703.700
Screw Cap Micro Tube DNA Low Binding, 1.5 ml, PP, with skirted base, with knurls, with assembled cap, with printed writing space and graduation, certified PCR Performance Tested (free of DNase/RNase, DNA and PCR inhibitors), 100 pcs./bag