Learn More
USA Scientific Inc 1.5 mL conical screw cap tube, amber
Catalog No. NC9184841
Encompass_Preferred
1.5 mL conical base screw cap microcentrifuge tube, graduated, amber with amber cap, 500/pack
Catalog No. NC9184841 Supplier USA Scientific Inc Supplier No. 14158707
1.5 mL conical base screw cap microcentrifuge tube, graduated, amber with amber cap, 500/pack