Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(5S)-4,5-dihydro-5-phenyl-1H-pyrazol-1-yl]-2,2-dimethyl-1-propanone | 2049868-46-2 | MFCD31813620 | 99.1% | 230.31 g·mol-1 | C14H18N2O | 50 MG
SDP

Catalog No. 5000246062
Encompass_Preferred

GSK'963 is a chiral, potent, and selective inhibitor of receptor-interacting protein kinase 1 (RIP1). It binds RIP1 with a reported FP-binding IC50 of about 29 nM and is used as a research tool to inhibit necroptosis in murine and human cell models. The compound has molecular formula C14H18N2O and molecular weight 230.31 g·mol-1.

  • Selective RIP1 kinase inhibitor (reported IC50 ~29 nM).
  • Used to inhibit necroptosis in murine and human cells in vitro.
  • Chiral small molecule; molecular formula C14H18N2O, MW 230.31 g·mol-1.
  • Available in small research pack sizes (commonly 50 mg).
  • Supplier-reported purity typically ≈99% (varies by vendor).

Catalog No. 50-002-46062 Supplier Medchemexpress LLC Supplier No. HY103028A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

GSK'963 is a chiral, potent, and selective inhibitor of receptor-interacting protein kinase 1 (RIP1). It binds RIP1 with a reported FP-binding IC50 of about 29 nM and is used as a research tool to inhibit necroptosis in murine and human cell models. The compound has molecular formula C14H18N2O and molecular weight 230.31 g·mol-1.

  • Selective RIP1 kinase inhibitor (reported IC50 ~29 nM).
  • Used to inhibit necroptosis in murine and human cells in vitro.
  • Chiral small molecule; molecular formula C14H18N2O, MW 230.31 g·mol-1.
  • Available in small research pack sizes (commonly 50 mg).
  • Supplier-reported purity typically ≈99% (varies by vendor).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.