Learn More
Fine Science Tools Ring Forceps, Stainless Steel, Straight, 8.5 cm
SDP
Catalog No. NC1325243
Encompass
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 1.5 x 0.45mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm
Catalog No. NC1325243 Supplier Fine Science Tools Supplier No. 1110109
Tip Shape: Straight/ Tips: Smooth/ Tip Dimensions: 1.5 x 0.45mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm