Learn More
Medchemexpress LLC 1-(5Z,8Z,11Z,14Z)-5,8,11,14-eicosatetraen-1-yl-1H-pyrrole-2,5-dione | 876305-42-9 | MFCD11521506 | 98.3% | 369.54 | C24H35NO2 | 5 MG
SDP
Catalog No. 5000221814
Encompass_Preferred
N-Arachidonyl maleimide is an irreversible inhibitor of monoacylglycerol lipase (MAGL) used in research to modulate endocannabinoid signaling by increasing 2-arachidonyl glycerol (2-AG) levels. It inhibits MAGL with an IC50 of 140 nM and has been characterized for preventing cerebellar membrane-mediated degradation of 2-AG. The compound is supplied in solid and solution formats for biochemical and cellular studies.
  • Irreversible MAGL inhibitor with reported IC50 of 140 nM.
  • Modulates 2-AG levels by inhibiting enzymatic degradation.
  • High reported purity (≈98.3%), suitable for research applications.
  • Available in multiple pack sizes and a pre-made solution for experimental flexibility.
  • Molecular formula C24H35NO2; molecular weight 369.54 g·mol⁻1.
  • Commonly used in biochemical assays to study endocannabinoid metabolism.
Catalog No. 50-002-21814 Supplier Medchemexpress LLC Supplier No. HY1365625MG
N-Arachidonyl maleimide is an irreversible inhibitor of monoacylglycerol lipase (MAGL) used in research to modulate endocannabinoid signaling by increasing 2-arachidonyl glycerol (2-AG) levels. It inhibits MAGL with an IC50 of 140 nM and has been characterized for preventing cerebellar membrane-mediated degradation of 2-AG. The compound is supplied in solid and solution formats for biochemical and cellular studies.
  • Irreversible MAGL inhibitor with reported IC50 of 140 nM.
  • Modulates 2-AG levels by inhibiting enzymatic degradation.
  • High reported purity (≈98.3%), suitable for research applications.
  • Available in multiple pack sizes and a pre-made solution for experimental flexibility.
  • Molecular formula C24H35NO2; molecular weight 369.54 g·mol⁻1.
  • Commonly used in biochemical assays to study endocannabinoid metabolism.