Learn More
Medchemexpress LLC 1-[(6,8-dimethyl-2-oxo-1H-quinolin-3-yl)methyl]-3-(4-ethoxyphenyl)-1-(2-hydroxyethyl)thiourea | 484006-66-8 | 98.2% | 425.54 g/mol | C23H27N3O3S | 5 MG
SDP
Catalog No. 5000246233
Encompass_Preferred
β-Glucuronidase-IN-1 is a potent, selective, uncompetitive inhibitor of E. coli bacterial β-glucuronidase used in biochemical and cellular research. It exhibits an IC50 of 283 nM and a Ki of 164 nM, and is supplied as a high-purity solid for assay development and inhibitor characterization.
  • Potent, selective uncompetitive inhibitor of E. coli β-glucuronidase.
  • Biochemical potency: IC50 283 nM; Ki 164 nM.
  • High purity solid suitable for research assays (≈98.2%).
  • Good solubility in DMSO (100 mg/mL) for convenient stock preparation.
  • Available as small-scale solid and as a DMSO solution for convenience.
  • Stable when stored as powder at -20°C (recommended storage conditions).
Catalog No. 50-002-46233 Supplier Medchemexpress LLC Supplier No. HY1030815MG
β-Glucuronidase-IN-1 is a potent, selective, uncompetitive inhibitor of E. coli bacterial β-glucuronidase used in biochemical and cellular research. It exhibits an IC50 of 283 nM and a Ki of 164 nM, and is supplied as a high-purity solid for assay development and inhibitor characterization.
  • Potent, selective uncompetitive inhibitor of E. coli β-glucuronidase.
  • Biochemical potency: IC50 283 nM; Ki 164 nM.
  • High purity solid suitable for research assays (≈98.2%).
  • Good solubility in DMSO (100 mg/mL) for convenient stock preparation.
  • Available as small-scale solid and as a DMSO solution for convenience.
  • Stable when stored as powder at -20°C (recommended storage conditions).