Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,6-anhydro-β-D-glucopyranose-d7 | 2483832-05-7 | 99.9% | 169.18 g/mol | C6H3D7O5 | 10 MG
SDP

Catalog No. 5000209777
Encompass_Preferred

Levoglucosan-d7 is a deuterium-labeled analogue of levoglucosan (1,6-anhydro-β-D-glucopyranose) used as a stable-isotope standard and tracer in analytical chemistry, particularly mass spectrometry and environmental analysis.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.9%) for accurate quantitation.
  • Available in small analytical pack sizes, such as 10 mg.
  • Solid powder format with defined storage conditions to preserve stability.
  • Molecular formula C6H3D7O5, molecular weight 169.18 g/mol.

Catalog No. 50-002-09777 Supplier Medchemexpress LLC Supplier No. HYW050145S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Levoglucosan-d7 is a deuterium-labeled analogue of levoglucosan (1,6-anhydro-β-D-glucopyranose) used as a stable-isotope standard and tracer in analytical chemistry, particularly mass spectrometry and environmental analysis.

  • Deuterium-labeled internal standard for mass spectrometry.
  • High purity (99.9%) for accurate quantitation.
  • Available in small analytical pack sizes, such as 10 mg.
  • Solid powder format with defined storage conditions to preserve stability.
  • Molecular formula C6H3D7O5, molecular weight 169.18 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.