Learn More
Medchemexpress LLC 1,6-anhydro-β-D-glucopyranose-d7 | 2483832-05-7 | 99.9% | 169.18 g/mol | C6H3D7O5 | 25 MG
SDP
Catalog No. 5000209078
Encompass_Preferred
Levoglucosan-d7 is the deuterium-labeled isotopic form of levoglucosan (1,6-anhydro-β-D-glucopyranose), supplied as an analytical standard for research and mass spectrometry applications.
  • Deuterium-labeled isotopic standard for levoglucosan.
  • High purity (99.9%).
  • Cas number 2483832-05-7.
  • Provided as a solid suitable for analytical use.
  • Common application: internal standard in mass spectrometry and tracer studies of biomass pyrolysis.
  • Available in small quantities for analytical workflows (for example, 25 mg).
Catalog No. 50-002-09078 Supplier Medchemexpress LLC Supplier No. 25MGHYW050145S
Levoglucosan-d7 is the deuterium-labeled isotopic form of levoglucosan (1,6-anhydro-β-D-glucopyranose), supplied as an analytical standard for research and mass spectrometry applications.
  • Deuterium-labeled isotopic standard for levoglucosan.
  • High purity (99.9%).
  • Cas number 2483832-05-7.
  • Provided as a solid suitable for analytical use.
  • Common application: internal standard in mass spectrometry and tracer studies of biomass pyrolysis.
  • Available in small quantities for analytical workflows (for example, 25 mg).