Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,6-anhydro-β-D-glucopyranose-d7 | 2483832-05-7 | 99.9% | 169.18 g/mol | C6H3D7O5 | 5 MG
SDP

Catalog No. 5000209914
Encompass_Preferred

Levoglucosan-d7 is the deuterium-labeled isotopologue of levoglucosan intended for research and analytical applications, commonly used as an internal standard in mass spectrometry assays.

  • deuterium-labeled levoglucosan (stable isotope standard)
  • intended for research and analytical applications, such as internal standard use in mass spectrometry
  • cas number 2483832-05-7
  • molecular formula C6H3D7O5
  • molecular weight 169.18 g/mol
  • purity 99.9%
  • package size 5 mg
  • storage: powder -20 °C (3 years), 4 °C (2 years); in solvent -80 °C (6 months), -20 °C (1 month)

Catalog No. 50-002-09914 Supplier Medchemexpress LLC Supplier No. HYW050145S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Levoglucosan-d7 is the deuterium-labeled isotopologue of levoglucosan intended for research and analytical applications, commonly used as an internal standard in mass spectrometry assays.

  • deuterium-labeled levoglucosan (stable isotope standard)
  • intended for research and analytical applications, such as internal standard use in mass spectrometry
  • cas number 2483832-05-7
  • molecular formula C6H3D7O5
  • molecular weight 169.18 g/mol
  • purity 99.9%
  • package size 5 mg
  • storage: powder -20 °C (3 years), 4 °C (2 years); in solvent -80 °C (6 months), -20 °C (1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.