Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,6-Anhydro-β-D-glucose, ≥99% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111014329 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4329 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4329 Supplier MilliporeSigma Supelco Supplier No. 0672425MG
Add to Cart
Add to Cart

Specifications

CAS 498-07-7
Linear Formula C6H10O5
MDL Number MFCD00063248
Melting Point 182°C to 184°C
Synonym Levoglucosan
Type Analytical Standard
Quantity 25 mg
Formula Weight 162.14
Percent Purity ≥99% (TLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.